missing translation for 'onlineSavingsMsg'
Learn More

1,2,2,6,6-Pentamethyl-4-piperidinol, 99%

Product Code. 10043420 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
1kg
250g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10043420 1 kg Plastic bottle 1kg
10058521 5 g Glass bottle 5g
10421301 250 g Plastic bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10043420 Supplier Thermo Scientific Acros Supplier No. 300860010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2403-89-6 | C10H21NO | 171.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2403-89-6
Molecular Formula C10H21NO
Molecular Weight (g/mol) 171.28
MDL Number MFCD00191220
InChI Key NWHNXXMYEICZAT-UHFFFAOYSA-N
Synonym 1,2,2,6,6-pentamethyl-4-piperidinol, 4-piperidinol, 1,2,2,6,6-pentamethyl, 4-hydroxy-1,2,2,6,6-pentamethylpiperidine, unii-lr520bam5v, 1,2,2,6,6-pentamethyl-4-hydroxypiperidine, lr520bam5v, methyl-taa-ol, hopemp, pubchem22251, acmc-209g8a
PubChem CID 75472
IUPAC Name 1,2,2,6,6-pentamethylpiperidin-4-ol
SMILES CC1(CC(CC(N1C)(C)C)O)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 70°C to 76°C
Color Yellow
Density 0.9670g/mL
Boiling Point 238°C
Flash Point 118°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 1 kg
Specific Gravity 0.967
Solubility Information Solubility in water: in water: 4.8% (20°C). Other solubilities: in ethanol: 55% (20°C),in acetone: 38% (20°C),in chloroform: 52% (20°C)
Viscosity 15.3 mPa.s (80°C)
Formula Weight 171.28
Percent Purity 99%
Physical Form Flakes
Chemical Name or Material 1,2,2,6,6-Pentamethyl-4-piperidinol, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.