missing translation for 'onlineSavingsMsg'
Learn More

1-[2-(2,5-Dimethyl-1H-pyrrol-1-yl)ethyl]piperazine, ≥90%, Thermo Scientific™

Product Code. 10386564
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10386564 10 g 10g
10613603 250 mg 250mg
10223032 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10386564 Supplier Thermo Scientific Maybridge Supplier No. MO00470EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 5059-36-9
Molecular Formula C12H21N3
Molecular Weight (g/mol) 207.32
MDL Number MFCD03407319
InChI Key AUSXFRFXXDHSTC-UHFFFAOYSA-N
PubChem CID 247631
IUPAC Name 1-[2-(2,5-dimethyl-1H-pyrrol-1-yl)ethyl]piperazine
SMILES CC1=CC=C(C)N1CCN1CCNCC1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 10 g
Synonym 1-2-2,5-dimethyl-1h-pyrrol-1-yl ethyl piperazine, 1-2-2,5-dimethyl-pyrrol-1-yl-ethyl-piperazine, 1-2-2,5-dimethylpyrrol-1-yl ethyl piperazine, piperazine,1-2-2,5-dimethyl-1h-pyrrol-1-yl ethyl, 2-2,5-dimethylpyrrolyl ethyl piperazine, enamine_004518, piperazine, 1-2-2,5-dimethyl-1h-pyrrol-1-yl ethyl
Formula Weight 207.32
Percent Purity 97%
Chemical Name or Material 1-[2-(2, 5-Dimethyl-1H-pyrrol-1-yl)ethyl]piperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.