missing translation for 'onlineSavingsMsg'
Learn More

1-(1H-Pyrrol-3-yl)ethan-1-one, 97%, Maybridge™

Product Code. 10416652
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10416652 10 g Amber Glass Bottle 10g
10131012 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10416652 Supplier Thermo Scientific Maybridge Supplier No. BTB07157EA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1072-82-8
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
MDL Number MFCD00067759
InChI Key KHHSXHXUQVNBGA-UHFFFAOYSA-N
Synonym 3-acetylpyrrole, 1-1h-pyrrol-3-yl ethanone, 1-1h-pyrrol-3-yl ethan-1-one, ethanone, 1-1h-pyrrol-3-yl, 1-1h-pyrrol-3-yl-ethanone, 3-acetyl-1h-pyrrole, 3-acetyl pyrrole, pubchem22508, 3-acetyl-1h-pyrroline
PubChem CID 2737793
IUPAC Name 1-(1H-pyrrol-3-yl)ethanone
SMILES CC(=O)C1=CNC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 109.13
Percent Purity 97%
Chemical Name or Material 1-(1H-pyrrol-3-yl)ethan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.