missing translation for 'onlineSavingsMsg'
Learn More

1,18-Octadecanedicarboxylic acid

Product Code. 11365017
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
250 g
Unit Size:
10g
250g
2g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11365017 50 g 50g
11375017 2 g 2g
11355017 10 g 10g
11345017 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11365017 Supplier Thermo Scientific Alfa Aesar Supplier No. 042004.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2424-92-2 | C20H38O4 | 342.52 g/mol

1,18-Octadecanedicarboxylic acid is used as an intermediate in organic synthesis, as dispersing agent, emulsifying agent, as lubricant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2424-92-2
Molecular Formula C20H38O4
Molecular Weight (g/mol) 342.52
MDL Number MFCD00059627
InChI Key JJOJFIHJIRWASH-UHFFFAOYSA-N
Synonym eicosanedioic acid, unii-sod235tgnz, 1,18-octadecanedicarboxylic acid, sod235tgnz, octadecane-1,18-dicarboxylic acid, eicosanedioicacid, 1,20-icosanedioic acid, 1,20-eicosanedioic acid, eicosa-1,20-dioic acid
PubChem CID 75502
ChEBI CHEBI:73728
IUPAC Name icosanedioic acid
SMILES C(CCCCCCCCCC(=O)O)CCCCCCCCC(=O)O

Specifications

Melting Point 127°C
Linear Formula HO2C(CH2)18CO2H
Quantity 50 g
Solubility Information Slightly soluble in water.
Formula Weight 342.52
Physical Form Powder
Chemical Name or Material 1,18-Octadecanedicarboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.