missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,16-Hexadecanedioic-d28 Acid, CDN
Specifications
Specifications
| Chemical Name or Material | Hexadecanedioic Acid-D28 |
| CAS | 130348-90-2 |
| Molecular Formula | C16 D28 H2 O4 |
| Quantity | 0.05 g |
| Synonym | Hexadecanedioic-d28 acid (9 CI), Hexadecanedioic acid-d28, 1,16-Hexadecanedioic-d28 acid, 1,14-Tetradecanedicarboxylic-d28 acid, Hexadecanedioic Acid-D28 |
| InChI Formula | InChI=1 S/C16H30O4/c17-15(18)13-11-9-7-5-3-1-2-4-6-8-10-12-14-16(19)20/h1-14H2,(H,17,18)(H,19,20)/i1D2,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2 |
| SMILES | [2 H]C([2 H])(C(=O)O)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)O |
| IUPAC Name | 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15-octacosadeuteriohexadecanedioic acid |
| Molecular Weight (g/mol) | 314.5795 |
| Formula Weight | 314.3902 g/mol |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?