missing translation for 'onlineSavingsMsg'
Learn More

1,12-Dibromododecane, 98%

Product Code. 11494777
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494777 25 g 25g
11484777 5 g 5g
11404787 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494777 Supplier Thermo Scientific Alfa Aesar Supplier No. B21254.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3344-70-5 | C12H24Br2 | 328.132 g/mol

1,12-Dibromododecane, is used as an organic chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,12-Dibromododecane, is used as an organic chemical intermediate.

Solubility
Soluble in hot methanol- almost transparency.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions.Stable. Combustible. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3344-70-5
Molecular Formula C12H24Br2
Molecular Weight (g/mol) 328.132
MDL Number MFCD00000226
InChI Key ZJJATABWMGVVRZ-UHFFFAOYSA-N
Synonym dodecamethylene dibromide, dodecane, 1,12-dibromo, alpha,omega-dibromododecane, labotest-bb ltbb002671, pubchem3879, 1, 12-dibromododecane, 1,12-dibromo-dodecane, 1,12-bis bromanyl dodecane, 4-01-00-00503 beilstein handbook reference, ksc225m2h
PubChem CID 18766
IUPAC Name 1,12-dibromododecane
SMILES C(CCCCCCBr)CCCCCBr

Specifications

Melting Point 38°C to 42°C
Boiling Point 215°C (15 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 1742763
Solubility Information Soluble in hot methanol- almost transparency.
Formula Weight 328.13
Percent Purity 98%
Chemical Name or Material 1,12-Dibromododecane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.