missing translation for 'onlineSavingsMsg'
Learn More

1,11b-Dedihydrotetrabenazine, TRC

Product Code. 30005127
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
50 mg
Unit Size:
10mg
2.5mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005127 10 mg 10mg
30009444 2.5 mg 2.5mg
30008649 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005127 Supplier Toronto Research Chemicals Supplier No. TRCD45360510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100322-43-8
Molecular Formula C19H25NO3
Molecular Weight (g/mol) 315.41
Synonym 3,4,6,7-Tetrahydro-9,10-dimethoxy-3-(2-methylpropyl)-2H-benzo[a]quinolizin-2-one, 3,4,6,7-Tetrahydro-3-isobutyl-9,10-dimethoxy-2H-benzo[a]quinolizin-2-one;
SMILES O=C1C(CC(C)C)CN2CCC3=CC(OC)=C(OC)C=C3C2=C1

Specifikationer

Quantity 10 mg
InChI Formula InChI=1S/C19H25NO3/c1-12(2)7-14-11-20-6-5-13-8-18(22-3)19(23-4)9-15(13)16(20)10-17(14)21/h8-10,12,14H,5-7,11H2,1-4H3
Formula Weight 315.1834
Chemical Name or Material 1,11b-Dedihydrotetrabenazine
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.