missing translation for 'onlineSavingsMsg'
Learn More

1,11-Undecanedicarboxylic Acid, TRC

Product Code. 30003233
Change view
Click to view available options
Quantity:
2.5 g
5 g
10 g
Unit Size:
10g
2.5g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30003233 10 g 10g
30012309 5 g 5g
30006199 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003233 Supplier Toronto Research Chemicals Supplier No. TRCU78766510G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 505-52-2
Molecular Formula C13H24O4
Molecular Weight (g/mol) 244.33
Synonym 1,13-Tridecanedioic Acid, Brassylic Acid, NSC 9498, α,ω-Tridecanedioic Acid, ω-Carboxylauric Acid;
IUPAC Name tridecanedioic acid
SMILES OC(=O)CCCCCCCCCCCC(=O)O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C13H24O4/c14-12(15)10-8-6-4-2-1-3-5-7-9-11-13(16)17/h1-11H2,(H,14,15)(H,16,17)
Formula Weight 244.1675
Chemical Name or Material 1,11-Undecanedicarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.