missing translation for 'onlineSavingsMsg'
Learn More

1,10-Phenanthroline, TRC

Product Code. 30426491
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30426491 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30426491 Supplier Toronto Research Chemicals Supplier No. TRCP29504025G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,10-Phenanthroline
CAS 66-71-7
Purity Grade Notes HPLC
Molecular Formula C12 H8 N2
Quantity 25 g
Synonym 1,10-o-Phenanthroline, 11,10-Phenanthroline, 4,5-Diazaphenanthrene, Activ-8, NSC 203545, o-Phenanthroline, TS 20, TS 20 (amine), β-Phenanthroline
InChI Formula InChI=1S/C12H8N2/c1-3-9-5-6-10-4-2-8-14-12(10)11(9)13-7-1/h1-8H
SMILES c1cnc2c(c1)ccc3cccnc23
IUPAC Name 1,10-phenanthroline
Molecular Weight (g/mol) 180.21
Formula Weight 180.0687
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.