missing translation for 'onlineSavingsMsg'
Learn More

1,10-Phenanthroline, Monohydrate, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 15528824 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
10 g
Packaging:
Wide Mouth Amber Glass Bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15528824 10 g Wide Mouth Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15528824 Supplier Avantor J.T.Baker Supplier No. T17002

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 5144-89-8
Packaging Wide Mouth Amber Glass Bottle
Molecular Formula C12H10N2O
MDL Number MFCD00149973
Quantity 10 g
Synonym 1,10-phenanthroline hydrate, 1,10-phenanthroline monohydrate, o-phenanthroline monohydrate, unii-ksx215x00e, 1,10-fenantrolina, 4,5-phenanthroline monohydrate, 1,10-phenanthroline, monohydrate, 1,10-fenanthrolin, 1,10-phenanthrolineo-phenanthroline, 1, 10-phenanthroline monohydrate
InChI Key PPQJCISYYXZCAE-UHFFFAOYSA-N
SMILES O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
IUPAC Name 1,10-phenanthroline;hydrate
Molecular Weight (g/mol) 198.23
PubChem CID 21226
Formula Weight 198.22
Grade BAKER ANALYZED™ ACS Reagent
Chemical Name or Material 1,10-Phenanthroline, Monohydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.