missing translation for 'onlineSavingsMsg'
Learn More

1,10-Phenanthroline iron(II) perchlorate, Thermo Scientific Chemicals

Product Code. 11357308
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357308 2 g 2g
11367308 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357308 Supplier Thermo Scientific Alfa Aesar Supplier No. 030557.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Oxidation, reduction indicator

1,10-Phenanthroline iron(II) perchlorate is used as an oxidation, reduction indicator. It is used for the detection, identification, analysis, of chemical, biological, or pathologic processes or conditions. Agents that are capable of inserting themselves between the successive bases in DNA, thus kinking, uncoiling or otherwise deforming it and therefore preventing its proper functioning. They are used in the study of DNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14586-54-0
Molecular Formula C36H24Cl2FeN6O8
Molecular Weight (g/mol) 795.367
MDL Number MFCD00050959
InChI Key QIZDTKUCQUZJHS-UHFFFAOYSA-L
Synonym 1,10-phenanthroline ferrous perchlorate, iron 2+ ; 1,10-phenanthroline; diperchlorate, iron 2+ perchlorate 1,10-phenanthroline 1:2:3, iron 2+ , tris 1,10-phenanthroline-.kappa.n1,.kappa.n10-, oc-6-11-, diperchlorate, tris 1,10-phenanthroline-n1,n10 iron diperchlorate, 3c12h8n2.2clo4.fe, iron 2+ , tris 1,10-phenanthroline-kappan1,kappan10-, oc-6-11-, diperchlorate, iron 2+ , tris 1,10-phenanthroline-kappan1,kappan10-, oc-6-11-, perchlorate 1:2, iron 2+ , tris 1,10-phenanthroline-.kappa.n1,.kappa.n10-, oc-6-11-, perchlorate 1:2
PubChem CID 84537
IUPAC Name iron(2+);1,10-phenanthroline;diperchlorate
SMILES C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Fe+2]

Specifications

Color Red
Quantity 2 g
UN Number UN1479
Formula Weight 795.37
Physical Form Crystalline
Chemical Name or Material 1,10-Phenanthroline iron(II) perchlorate
compliance-icons compliance-icons
Product Identifier
  • 1,10-Phenanthroline iron(II) perchlorate
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.