missing translation for 'onlineSavingsMsg'
Learn More

1,10-Phenanthroline Ferrous Solution 0.025M, Pure, Fisher Chemical

Product Code. 10244490 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10244490 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10244490 Supplier Fisher Chemical Supplier No. P/2220/PB05

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Packaging Plastic Coated Glass bottle
CAS 14634-91-4
Physical Form Liquid
Molecular Formula C36H24FeN6O4S
MDL Number MFCD00036428
Color Red
InChI Key CIWXFRVOSDNDJZ-UHFFFAOYSA-L
SMILES [Fe++].[O-]S([O-])(=O)=O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
IUPAC Name λ²-iron(2+) tris(1,10-phenanthroline) sulfate
Molecular Weight (g/mol) 692.53
PubChem CID 84567
Quantity 100 mL
Formula Weight 692.53g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.