missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,10-Diiododecane, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
1,10-Diiododecane is used as an alkylating agent in the synthesis of symmetrical and unsymmetrical bis-cryptophanes and as a crosslinking reagent for tetrabutylammonium polygalacturonic acid. it is also used to produce decane at temperature of 20°C.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,10-Diiododecane is used as an alkylating agent in the synthesis of symmetrical and unsymmetrical bis-cryptophanes and as a crosslinking reagent for tetrabutylammonium polygalacturonic acid. it is also used to produce decane at temperature of 20°C.
Solubility
Soluble in methanol.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to light.
1,10-Diiododecane is used as an alkylating agent in the synthesis of symmetrical and unsymmetrical bis-cryptophanes and as a crosslinking reagent for tetrabutylammonium polygalacturonic acid. it is also used to produce decane at temperature of 20°C.
Solubility
Soluble in methanol.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to light.
Specifications
Melting Point | 28°C to 32°C |
Density | 2.35 |
Boiling Point | 197°C to 200°C (12 mmHg) |
Flash Point | >110°C (230°F) |
Quantity | 10 g |
Beilstein | 1738614 |
Sensitivity | Light sensitive |
Synonym | decamethylene diiodide, decane, 1,10-diiodo, 1,10-diiododecan, pubchem3844, acmc-1btxk, decane,1,10-diiodo, 1,10-diiododecane |
Solubility Information | Soluble in methanol. |
Formula Weight | 394.08 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'