missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,10-Dibromodecane, 97%
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
Description
1,10-Dibromodecane is used as an alkylating agent and as synthetic organic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,10-Dibromodecane is used as an alkylating agent and as synthetic organic intermediate.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sea
1,10-Dibromodecane is used as an alkylating agent and as synthetic organic intermediate.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sea
Specifications
Specifications
| Melting Point | 26°C to 30°C |
| Density | 1.34 |
| Boiling Point | 157°C to 159°C (14 mmHg) |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.493 |
| Quantity | 100 g |
| Beilstein | 506156 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 300.08 |
| Percent Purity | 97% |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction