missing translation for 'onlineSavingsMsg'
Learn More

1,10-Diaminodecane, TRC

Product Code. 30378933
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30378933 500 mg 500mg
30376852 250 mg -
30376541 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30378933 Supplier Toronto Research Chemicals Supplier No. TRCD493628500MG
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 646-25-3
Molecular Formula C10H24N2
Molecular Weight (g/mol) 172.31
Synonym 1,10-Decamethylenediamine, 1,10-Decylenediamine, 1,10-Diamino-n-decane, 1,10-Diaminodecane, 1,6-Decanediamine, Decamethylenediamine, Fentamine HP 102, NCI 10400, NSC 10400
IUPAC Name decane-1,10-diamine
SMILES NCCCCCCCCCCN

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C10H24N2/c11-9-7-5-3-1-2-4-6-8-10-12/h1-12H2
Formula Weight 172.1939
Chemical Name or Material 1,10-Diaminodecane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.