missing translation for 'onlineSavingsMsg'
Learn More

1,10-Decanediol, 99%

Product Code. 10572971 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 10572971

Brand: Thermo Scientific Acros 181041000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-47-0 | C10H22O2 | 174.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-47-0
Molecular Formula C10H22O2
Molecular Weight (g/mol) 174.28
MDL Number MFCD00004749
InChI Key FOTKYAAJKYLFFN-UHFFFAOYSA-N
Synonym 1,10-decanediol, decamethylene glycol, decamethylenediol, 1,10-dihydroxydecane, 1,10-decamethylenediol, 1,10-decamethylene diol, 1,6-bis 2-hydroxyethyl hexane, alpha,omega-decanediol, unii-5i577udk52, 1,10-ddecanediol
PubChem CID 37153
IUPAC Name decane-1,10-diol
SMILES C(CCCCCO)CCCCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 71°C to 75°C
Color White
Boiling Point 297°C
Flash Point 152°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula HO(CH2)10OH
Quantity 100 g
Beilstein 01, II, 560
Merck Index 15, 2853
Solubility Information Solubility in water: insoluble in water. Other solubilities: freely soluble in alcohol and warm ether,practically insoluble in petroleum ether
Formula Weight 174.28
Percent Purity 99%
Physical Form Powder, Flakes or Pellets
Chemical Name or Material 1, 10-Decanediol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.