missing translation for 'onlineSavingsMsg'
Learn More

1,10-Decanediol, 97%

Product Code. 11421648
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11421648 500 g 500g
11411648 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11421648 Supplier Thermo Scientific Alfa Aesar Supplier No. A13923.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 112-47-0 | C10H22O2 | 174.284 g/mol

1,10-Decanediol is an minor intermediate in the production of polyesters. It also used in essence, perfumes and in pharmaceuticals. The diacrylate is hydrophobic and used for flexible dental materials, radiation curable coatings, and for lenses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,10-Decanediol is an minor intermediate in the production of polyesters. It also used in essence, perfumes and in pharmaceuticals. The diacrylate is hydrophobic and used for flexible dental materials, radiation curable coatings, and for lenses.

Solubility
Soluble in alcohol. Insoluble in water, petroleum ether.

Notes
Store in a cool, dry, well-ventilated area. Incompatible with oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-47-0
Molecular Formula C10H22O2
Molecular Weight (g/mol) 174.284
MDL Number MFCD00004749
InChI Key FOTKYAAJKYLFFN-UHFFFAOYSA-N
Synonym 1,10-decanediol, decamethylene glycol, decamethylenediol, 1,10-dihydroxydecane, 1,10-decamethylenediol, 1,10-decamethylene diol, 1,6-bis 2-hydroxyethyl hexane, alpha,omega-decanediol, unii-5i577udk52, 1,10-ddecanediol
PubChem CID 37153
IUPAC Name decane-1,10-diol
SMILES C(CCCCCO)CCCCO

Specifications

Melting Point 71°C to 74°C
Density 1.08
Boiling Point 297°C
Flash Point 152°C (306°F)
Quantity 500 g
Beilstein 1698975
Merck Index 14,2849
Solubility Information Soluble in alcohol. Insoluble in water,petroleum ether.
Formula Weight 174.28
Percent Purity 97%
Chemical Name or Material 1,10-Decanediol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.