missing translation for 'onlineSavingsMsg'
Learn More

1,10-Decanedicarboxylic acid, 99%

Product Code. p-3757382 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
500g
5g
This item is not returnable. View return policy

Product Code. 10294130

Brand: Thermo Scientific Acros 111880050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 693-23-2 | C12H22O4 | 230.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 693-23-2
Molecular Formula C12H22O4
Molecular Weight (g/mol) 230.3
MDL Number MFCD00002735
InChI Key TVIDDXQYHWJXFK-UHFFFAOYSA-N
Synonym 1,12-dodecanedioic acid, 1,10-decanedicarboxylic acid, decamethylenedicarboxylic acid, 1,10-dicarboxydecane, n-dodecanedioic acid, unii-978yu42q6i, dsstox_cid_7297, dsstox_rid_78394, dsstox_gsid_27297, n-dodecanedioate
PubChem CID 12736
ChEBI CHEBI:4676
IUPAC Name dodecanedioic acid
SMILES C(CCCCCC(=O)O)CCCCC(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 125°C to 130°C
Color White
Boiling Point 245°C (10.0 mmHg)
Flash Point 220°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula HO2C(CH2)10CO2H
Quantity 5 g
Beilstein 02, IV, 2126
Solubility Information Solubility in water: < 0.1g/L (20 c)
Viscosity 8.4 mPa.s (140°C)
Formula Weight 230.3
Percent Purity 99%
Physical Form Pellets or Powder
Chemical Name or Material 1, 10-Decanedicarboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 1,10-Decanedicarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.