missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Thiocarbonyldiimidazole, tech 90%

Product Code. 11489487
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11489487 250 g 250g
11469487 10 g 10g
11479487 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11489487 Supplier Thermo Scientific Alfa Aesar Supplier No. B22008.30
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6160-65-2 | C7H6N4S | 178.21 g/mol

1,1'-Thiocarbonyldiimidazole is used as a reagent for the deoxygenation of alcohols with tributyltin hydride and diols with lithium aluminum hydride. It is also used to deoxygenate carboxylic monosaccharide analogues. Further, it is a useful reagent involved in biochemical synthesis. In addition to this, it is used to prepare 1-[bis-(4-methoxy-phenyl)-methyl]-1H-imidazole by reacting with 4,4'-dimethoxy-benzhydrol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1′-Thiocarbonyldiimidazole is used as a reagent for the deoxygenation of alcohols with tributyltin hydride and diols with lithium aluminum hydride. It is also used to deoxygenate carboxylic monosaccharide analogues. Further, it is a useful reagent involved in biochemical synthesis. In addition to this, it is used to prepare 1-[bis-(4-methoxy-phenyl)-methyl]-1H-imidazole by reacting with 4,4′-dimethoxy-benzhydrol.

Solubility
Soluble in terahydrofuran, toluene and dichloromethane.

Notes
Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Store in a cool place. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6160-65-2
Molecular Formula C7H6N4S
Molecular Weight (g/mol) 178.21
MDL Number MFCD00005289
InChI Key RAFNCPHFRHZCPS-UHFFFAOYSA-N
Synonym 1,1'-thiocarbonyldiimidazole, thiocarbonyldiimidazole, n,n'-thiocarbonyldiimidazole, di 1h-imidazol-1-yl methanethione, 1,1-thiocarbonyldiimidazole, di imidazol-1-yl methanethione, n,n'-thiocarbonyldiimidazole tcdi, 1h-imidazole, 1,1'-carbonothioylbis, n,n-thiocarbonyldiimidazole, 1,1'-carbonothioylbis-1h-imidazole
PubChem CID 80264
IUPAC Name di(imidazol-1-yl)methanethione
SMILES S=C(N1C=CN=C1)N1C=CN=C1

Specifications

Melting Point 98°C to 102°C
Assay Percent Range 0.95
Quantity 250 g
Beilstein 609349
Sensitivity Moisture sensitive
Solubility Information Soluble in terahydrofuran,toluene and dichloromethane.
Formula Weight 178.22
Percent Purity 90%
Grade Technical
Chemical Name or Material 1,1'-Thiocarbonyldiimidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.