missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Thiocarbonyldiimidazole, 90%, Tech.

Product Code. 15750819 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15750819 25 g Glass bottle 25g
10686372 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15750819 Supplier Thermo Scientific Acros Supplier No. 168480250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6160-65-2 | C7H6N4S | 178.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 6160-65-2
Molecular Formula C7H6N4S
Molecular Weight (g/mol) 178.21
MDL Number MFCD00005289
InChI Key RAFNCPHFRHZCPS-UHFFFAOYSA-N
Synonym 1,1'-thiocarbonyldiimidazole, thiocarbonyldiimidazole, n,n'-thiocarbonyldiimidazole, di 1h-imidazol-1-yl methanethione, 1,1-thiocarbonyldiimidazole, di imidazol-1-yl methanethione, n,n'-thiocarbonyldiimidazole tcdi, 1h-imidazole, 1,1'-carbonothioylbis, n,n-thiocarbonyldiimidazole, 1,1'-carbonothioylbis-1h-imidazole
PubChem CID 80264
IUPAC Name di(imidazol-1-yl)methanethione
SMILES S=C(N1C=CN=C1)N1C=CN=C1

Specifica

CAS Min % 88.0
CAS Max % 100.0
Melting Point 96.0°C to 103.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Quantity 25 g
Fieser 01,1151; 02,411; 05,661; 06,583; 13,318
Solubility Information Solubility in water: decomposes.
Formula Weight 178.21
Percent Purity 90%
Grade Technical
Physical Form Powder
Chemical Name or Material 1, 1'-Thiocarbonyldiimidazole
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.