missing translation for 'onlineSavingsMsg'
Learn More

1-(1-Pyrenyl)ethanol, 98%, Thermo Scientific™

Product Code. 15244764 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15244764 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15244764 Supplier Thermo Scientific Acros Supplier No. 449290010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 65954-42-9
Melting Point 111.0°C to 113.0°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C18H14O
Quantity 1 g
Synonym 1-1-pyrenyl ethanol, 1-hydroxyethylpyrene, alpha-methyl-1-pyrenemethanol, 1-1-hydroxyethyl pyrene, ccris 7293, 1-pyrenemethanol, alpha-methyl, 1-pyren-1-yl ethanol, 1-pyrenylethan-1-ol, 4-06-00-05012 beilstein handbook reference
Solubility Information Solubility in water: insoluble
InChI Key LFCWFVPSUMBNQM-UHFFFAOYSA-N
SMILES CC(C1=C2C=CC3=CC=CC4=C3C2=C(C=C4)C=C1)O
IUPAC Name 1-pyren-1-ylethanol
Molecular Weight (g/mol) 246.31
PubChem CID 119118
Formula Weight 246.31
Percent Purity 98%
Chemical Name or Material 1-(1-Pyrenyl)ethanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.