missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Oxybis(3,3,3-trifluoro-1-propanol) hemihydrate, 96%, Thermo Scientific™

Product Code. 15449265
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15449265 5 g 5g
15439265 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15449265 Supplier Thermo Scientific Alfa Aesar Supplier No. H51690.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 936107-87-8
Molecular Formula C6H8F6O3
Molecular Weight (g/mol) 242.12
MDL Number MFCD22989348
InChI Key MBQFJCIIFZJMTG-UHFFFAOYNA-N
Synonym 3,3,3-trifluoropropionaldehyde hemihydrate, 1,1'-oxybis 3,3,3-trifluoro-1-propanol hemihydrate
PubChem CID 16222109
IUPAC Name 3,3,3-trifluoro-1-(3,3,3-trifluoro-1-hydroxypropoxy)propan-1-ol
SMILES OC(CC(F)(F)F)OC(O)CC(F)(F)F

Specifications

Linear Formula [CF3CH2CH(OH)]2O·0.5H2O
Quantity 5 g
UN Number UN1988
Formula Weight 251.12 (242.12 Anhydrous)
Percent Purity 96%
Chemical Name or Material 1,1'-Oxybis(3,3,3-trifluoro-1-propanol) hemihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.