missing translation for 'onlineSavingsMsg'
Learn More

1-(1-Naphthyl)piperazine hydrochloride, Thermo Scientific Chemicals

Product Code. 15454539
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15454539 1 g 1g
15464539 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15454539 Supplier Thermo Scientific Alfa Aesar Supplier No. J62097.03
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 104113-71-5 | Molecular Formula: C14H17ClN2 | Molecular Weight: 248.754 g/mol

5-HT1 serotonin receptor agonist. 5-HT2 serotonin receptor antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • 5-HT1 serotonin receptor agonist
  • 5-HT2 serotonin receptor antagonist
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104113-71-5
Molecular Formula C14H17ClN2
Molecular Weight (g/mol) 248.754
MDL Number MFCD00055161
InChI Key ZYVYPNZFOCZLEM-UHFFFAOYSA-N
Synonym 1-1-naphthyl piperazine hydrochloride, 1-naphthalen-1-yl piperazine hydrochloride, naphthylpiperazine hydrochloride, 1-naphthalen-1-ylpiperazine hydrochloride, 1-naphthalen-1-yl-piperazine hydrochloride, piperazine,1-1-naphthalenyl, zlchem 1062, 1-1-naphthyl piperazine hcl, 1-naphthalen-1-yl-piperazine hcl salt, n-1-naphthyl piperazine hydrochloride
PubChem CID 2737391
IUPAC Name 1-naphthalen-1-ylpiperazine;hydrochloride
SMILES C1CN(CCN1)C2=CC=CC3=CC=CC=C32.Cl

Specifications

Color Purple
Quantity 1 g
Formula Weight 248.75
Physical Form Solid
Chemical Name or Material 1-(1-Naphthyl)piperazine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.