missing translation for 'onlineSavingsMsg'
Learn More

1-(1-Naphthyl)-2-(trimethylsilyl)acetylene, 97%, Thermo Scientific Chemicals

Product Code. 15452358
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104784-51-2
Molecular Formula C15H16Si
Molecular Weight (g/mol) 224.378
MDL Number MFCD04039886
InChI Key WATBCTJRCLNXNF-UHFFFAOYSA-N
Synonym 1-1-naphthyl-2-trimethylsilyl acetylene, trimethyl 2-naphthalen-1-ylethynyl silane, trimethyl 2-naphthalen-1-yl ethynyl silane, 1-naphthylethynyl trimethylsilane, 1-trimethylsilylethynyl naphthalene, 1-trimethylsilyl ethynyl naphthalene, trimethyl naphthalen-1-ylethynyl silane, trimethyl naphthalen-1-yl ethynyl silane, trimethylnaphthalen-1-ylethynylsilane
PubChem CID 4438232
IUPAC Name trimethyl(2-naphthalen-1-ylethynyl)silane
SMILES C[Si](C)(C)C#CC1=CC=CC2=CC=CC=C21

Specifications

Melting Point 41°C to 43°C
Density 0.968 g/mL
Boiling Point 292°C to 293°C
Flash Point >110°C (230°F)
Refractive Index 1.595
Quantity 25 g
Formula Weight 224.38
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(1-Naphthyl)-2-(trimethylsilyl)acetylene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'