missing translation for 'onlineSavingsMsg'
Learn More

1-(1-Naphthyl)-2-thiourea 97%, Thermo Scientific™

Product Code. 10229410 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass Bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10229410 25 g Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10229410 Supplier Thermo Scientific Acros Supplier No. 415430250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 86-88-4
CAS Min % 96.0
CAS Max % 100.0
Color Beige-Brown to Gray
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Molecular Formula C11H10N2S
MDL Number MFCD00041824
Quantity 25 g
Merck Index 15, 708
Synonym 1-1-naphthyl-2-thiourea, antu, 1-naphthylthiourea, dirax, anturat, rattrack, smeesana, alrato, bantu, kripid
InChI Key PIVQQUNOTICCSA-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C=CC=C2NC(=S)N
IUPAC Name naphthalen-1-ylthiourea
Molecular Weight (g/mol) 202.275
PubChem CID 736366
ChEBI CHEBI:82256
Formula Weight 202.28
Percent Purity ≥96.0%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 1-(1-Naphthyl)-2-thiourea, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-(1-Naphthyl)-2-thiourea
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.