missing translation for 'onlineSavingsMsg'
Learn More

1,1'-[Methylenebis(4,1-phenyleneoxy)]bis[3-[(1-methylethyl)amino]-2-propanol, TRC

Product Code. 30135893
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135893 50 mg 50mg
30134669 25 mg 25mg
17921575 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135893 Supplier Toronto Research Chemicals Supplier No. TRCM33155550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1225195-70-9
Molecular Formula C25 H38 N2 O4
Molecular Weight (g/mol) 430.58
Synonym (RS)-1-[4-[4-(2-Hydroxy-3-isopropylaminopropoxy)benzyl]phenoxy]-3-isopropylaminopropan-2-ol, Bisoprolol Fumarate Imp. C (EP), 2-Propanol, 1,1'-[methylenebis(4,1-phenyleneoxy)]bis[3-[(1-methylethyl)amino]- (ACI), 1,1'-[Methylenebis(4,1-phenyleneoxy)]bis[3-[(1-methylethyl)amino]-2-propanol] (ACI)
IUPAC Name 1-[4-[[4-[2-hydroxy-3-(propan-2-ylamino)propoxy]phenyl]methyl]phenoxy]-3-(propan-2-ylamino)propan-2-ol
SMILES CC(C)NCC(O)COc1ccc(Cc2ccc(OCC(O)CNC(C)C)cc2)cc1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C25H38N2O4/c1-18(2)26-14-22(28)16-30-24-9-5-20(6-10-24)13-21-7-11-25(12-8-21)31-17-23(29)15-27-19(3)4/h5-12,18-19,22-23,26-29H,13-17H2,1-4H3
Formula Weight 430.2832
Chemical Name or Material (RS)-1-[4-[4-(2-Hydroxy-3-isopropylaminopropoxy)benzyl]phenoxy]-3-isopropylaminopropan-2-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.