missing translation for 'onlineSavingsMsg'
Learn More

1-(1-Methyl-4-piperidinyl)piperazine, 98%

Product Code. 11416570
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11416570 1 g 1g
11426570 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11416570 Supplier Thermo Scientific Alfa Aesar Supplier No. H27624.03
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 23995-88-2 | C10H21N3 | 183.299 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23995-88-2
Molecular Formula C10H21N3
Molecular Weight (g/mol) 183.299
MDL Number MFCD01075186
InChI Key OHUMKYGINIODOY-UHFFFAOYSA-N
Synonym 1-1-methyl-4-piperidinyl piperazine, 1-1-methylpiperidin-4-yl piperazine, 1-n-methylpiperidin-4-yl piperazine, 1-1-methyl-piperidin-4-yl-piperazine, piperazine, 1-1-methyl-4-piperidinyl, 1-1-methyl-4-piperidyl piperazine, 1-1-methyl-4-piperidino piperazine, zlchem 509, acmc-1cq7x, 1-methyl-4-piperidyl piperazine
PubChem CID 566324
IUPAC Name 1-(1-methylpiperidin-4-yl)piperazine
SMILES CN1CCC(CC1)N2CCNCC2

Specifications

Melting Point 29°C to 32°C
Refractive Index 1.508
Quantity 1 g
UN Number UN3259
Sensitivity Air Sensitive
Formula Weight 183.3
Percent Purity 98%
Chemical Name or Material 1-(1-Methyl-4-piperidinyl)piperazine
compliance-icons compliance-icons
Product Identifier
  • 1-(1-Methyl-4-piperidinyl)piperazine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 1-(1-Methyl-4-piperidinyl)piperazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.