missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Ferrocenedimethanol, 98%, Thermo Scientific Chemicals

Product Code. 10402873 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10402873 1 g 1g
10280453 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10402873 Supplier Thermo Scientific Acros Supplier No. 382250010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1291-48-1
Molecular Formula C12H14FeO2
Molecular Weight (g/mol) 246.09
MDL Number MFCD02091688
Synonym ferrocene derivatives, 1,1'-bis hydroxymethyl ferrocen, 1,1'-ferrocenedimethanol, 1,1 inverted exclamation marka-ferrocenedimethanol
SMILES [Fe].OCc1cccc1.OCc1cccc1

Specifications

Melting Point 106.0°C to 109.0°C
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble.
Formula Weight 246.09
Percent Purity 98%
Chemical Name or Material 1,1′-Ferrocenedimethanol
compliance-icons
Product Identifier
  • 1,1'-Ferrocenedimethanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.