missing translation for 'onlineSavingsMsg'
Learn More

1,1-Diphenylethylene oxide

Product Code. 10095493 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10095493 5 g Glass bottle 5g
10782284 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10095493 Supplier Thermo Scientific Acros Supplier No. 391030050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 882-59-7 | C14H12O | 196.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 882-59-7
Molecular Formula C14H12O
Molecular Weight (g/mol) 196.25
MDL Number MFCD00040725
InChI Key PRLJMHVNHLTQJJ-UHFFFAOYSA-N
Synonym 1,1-diphenylethylene oxide, oxirane, 2,2-diphenyl, diphenyloxirane, 2,2-diphenyl-oxirane, 2,2-diphenyl oxirane, #, 1,1-diphenyl-ethylenoxide, 1,1-diphenyl-ethylene oxide, 1,1-diphenyl-1,2-epoxyethane
PubChem CID 93564
IUPAC Name 2,2-diphenyloxirane
SMILES C1C(O1)(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 54°C to 56°C
Color White
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5 g
Formula Weight 196.25
Percent Purity 96%
Physical Form Crystals
Chemical Name or Material 1, 1-Diphenylethylene oxide
compliance-icons compliance-icons
Product Identifier
  • 1,1-Diphenylethylene oxide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.