missing translation for 'onlineSavingsMsg'
Learn More

1,1-Diphenylethylene, 98%, Thermo Scientific Chemicals

Catalog Number 10675342
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
530-48-3
C14H12
180.25
ZMYIIHDQURVDRB-UHFFFAOYSA-N
1,1-diphenylethylene, ethene-1,1-diyldibenzene, 1,1-diphenylethene, benzene, 1,1'-ethenylidenebis, 1-phenylethenyl benzene, unii-bx0l5b6lll, as-diphenylethylene, .alpha.-phenylstyrene, 1-phenylvinyl benzene, bx0l5b6lll
10740
1-phenylethenylbenzene
C=C(C1=CC=CC=C1)C2=CC=CC=C2
This item is not returnable. View return policy
530-48-3
C14H12
180.25
ZMYIIHDQURVDRB-UHFFFAOYSA-N
1,1-diphenylethylene, ethene-1,1-diyldibenzene, 1,1-diphenylethene, benzene, 1,1'-ethenylidenebis, 1-phenylethenyl benzene, unii-bx0l5b6lll, as-diphenylethylene, .alpha.-phenylstyrene, 1-phenylvinyl benzene, bx0l5b6lll
10740
1-phenylethenylbenzene
C=C(C1=CC=CC=C1)C2=CC=CC=C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 6.0°C
Color Colorless
Density 1.0200g/mL
Boiling Point 270.0°C to 271.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (C6H5)2C=CH2
Refractive Index 1.6065 to 1.6085
Quantity 25 g
Beilstein 05,639
Merck Index 15,3356
Specific Gravity 1.02
Formula Weight 180.25
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1, 1-Diphenylethylene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'