missing translation for 'onlineSavingsMsg'
Learn More

1,1-Diphenyl-2-propyn-1-ol, 98%, Thermo Scientific Chemicals

Catalog Number 10414895
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
3923-52-2
C15H12O
208.26
MFCD00041570
SMCLTAARQYTXLW-UHFFFAOYSA-N
1,1-diphenyl-2-propyn-1-ol, diphenylethinylcarbinol, ethynyldiphenylmethanol, 1,1-diphenylpropargyl alcohol, diphenylethynylcarbinol, 3,3-diphenyl-3-hydroxy-1-propyne, benzenemethanol, .alpha.-ethynyl-.alpha.-phenyl, 2-propyn-1-ol, 1,1-diphenyl, benzenemethanol, alpha-ethynyl-alpha-phenyl, benzenemethanol, a-ethynyl-a-phenyl
92976
1,1-diphenylprop-2-yn-1-ol
OC(C#C)(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
3923-52-2
C15H12O
208.26
MFCD00041570
SMCLTAARQYTXLW-UHFFFAOYSA-N
1,1-diphenyl-2-propyn-1-ol, diphenylethinylcarbinol, ethynyldiphenylmethanol, 1,1-diphenylpropargyl alcohol, diphenylethynylcarbinol, 3,3-diphenyl-3-hydroxy-1-propyne, benzenemethanol, .alpha.-ethynyl-.alpha.-phenyl, 2-propyn-1-ol, 1,1-diphenyl, benzenemethanol, alpha-ethynyl-alpha-phenyl, benzenemethanol, a-ethynyl-a-phenyl
92976
1,1-diphenylprop-2-yn-1-ol
OC(C#C)(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 46.0°C to 50.0°C
Color White to Yellow
Boiling Point 183.0°C (20.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble.
Formula Weight 208.26
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material 1, 1-Diphenyl-2-propyn-1-ol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'