missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dioxobenzo[b]thiophen-2-ylmethyl N-succimidyl carbonate, 95%, Thermo Scientific™

Product Code. p-7047770
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11325585

Brand: Thermo Scientific Alfa Aesar L19733.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 197244-91-0
Molecular Formula C14H11NO7S
Molecular Weight (g/mol) 337.302
MDL Number MFCD03095506
InChI Key XBVMGLSMCWWRLS-UHFFFAOYSA-N
Synonym 1,1-dioxidobenzo b thiophen-2-yl methyl 2,5-dioxopyrrolidin-1-yl carbonate, bsmoc-osu, 1,1-dioxobenzo b thiophen-2-ylmethyl n-succinimidyl carbonate, 1,1-dioxobenzo b thiophen-2-ylmethyl n-succimidyl carbonate, 1,1-dioxo-1??-benzothiophen-2-yl methyl 2,5-dioxopyrrolidin-1-yl carbonate, n-benzo b thiophenesulfone-2-methoxycarbonyloxy succinimide, 2,5-dioxoazolidinyl 1,1-dioxobenzo d thiolen-2-yl methoxy formate, 1,1-dioxo-1-benzothiophen-2-yl methyl 2,5-dioxopyrrolidin-1-yl carbonate, 2-2,5-dioxopyrrolizinooxy carbonyloxy methyl benzo b thiophene 1,1-dioxide, carbonic acid, 1,1-dioxidobenzo b thien-2-yl methyl 2,5-dioxo-1-pyrrolidinyl ester
PubChem CID 3344958
IUPAC Name (1,1-dioxo-1-benzothiophen-2-yl)methyl (2,5-dioxopyrrolidin-1-yl) carbonate
SMILES C1CC(=O)N(C1=O)OC(=O)OCC2=CC3=CC=CC=C3S2(=O)=O

Specifications

Melting Point 140°C (decomposition)
Quantity 1 g
Beilstein 8274967
Sensitivity Moisture sensitive
Formula Weight 337.31
Percent Purity 95%
Chemical Name or Material 1,1-Dioxobenzo[b]thiophen-2-ylmethyl N-succimidyl carbonate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.