missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dioxobenzo[b]thiophen-2-ylmethyl N-succimidyl carbonate, 95%, Thermo Scientific™

Product Code. 11335585
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11335585 5 g 5g
11325585 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11335585 Supplier Thermo Scientific Alfa Aesar Supplier No. L19733.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 197244-91-0
Molecular Formula C14H11NO7S
Molecular Weight (g/mol) 337.302
MDL Number MFCD03095506
InChI Key XBVMGLSMCWWRLS-UHFFFAOYSA-N
Synonym 1,1-dioxidobenzo b thiophen-2-yl methyl 2,5-dioxopyrrolidin-1-yl carbonate, bsmoc-osu, 1,1-dioxobenzo b thiophen-2-ylmethyl n-succinimidyl carbonate, 1,1-dioxobenzo b thiophen-2-ylmethyl n-succimidyl carbonate, 1,1-dioxo-1??-benzothiophen-2-yl methyl 2,5-dioxopyrrolidin-1-yl carbonate, n-benzo b thiophenesulfone-2-methoxycarbonyloxy succinimide, 2,5-dioxoazolidinyl 1,1-dioxobenzo d thiolen-2-yl methoxy formate, 1,1-dioxo-1-benzothiophen-2-yl methyl 2,5-dioxopyrrolidin-1-yl carbonate, 2-2,5-dioxopyrrolizinooxy carbonyloxy methyl benzo b thiophene 1,1-dioxide, carbonic acid, 1,1-dioxidobenzo b thien-2-yl methyl 2,5-dioxo-1-pyrrolidinyl ester
PubChem CID 3344958
IUPAC Name (1,1-dioxo-1-benzothiophen-2-yl)methyl (2,5-dioxopyrrolidin-1-yl) carbonate
SMILES C1CC(=O)N(C1=O)OC(=O)OCC2=CC3=CC=CC=C3S2(=O)=O

Specifications

Melting Point 140°C (decomposition)
Quantity 5 g
Beilstein 8274967
Sensitivity Moisture sensitive
Formula Weight 337.31
Percent Purity 95%
Chemical Name or Material 1,1-Dioxobenzo[b]thiophen-2-ylmethyl N-succimidyl carbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.