missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dimethylpropargylamine, 95%

Product Code. 10289363 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10289363 25 g Glass bottle 25g
10351004 1 g Glass bottle 1g
10714344 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10289363 Supplier Thermo Scientific Acros Supplier No. 175620250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2978-58-7 | C5H9N | 83.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2978-58-7
Molecular Formula C5H9N
Molecular Weight (g/mol) 83.13
MDL Number MFCD00008052
InChI Key VUGCBIWQHSRQBZ-UHFFFAOYSA-N
Synonym 1,1-dimethylpropargylamine, 3-amino-3-methyl-1-butyne, 3-butyn-2-amine, 2-methyl, 1,1-dimethylprop-2-ynylamine, 2-methyl-3-butyn-2-amine, 1,1-dimethyl-prop-2-ynylamine, 1,1-dimethylpropynylamine, 1,1-dimethyl-2-propynylamine, 1,1-dimethylprop-3-ynylamine, 3-amino-3-methylbutyne
PubChem CID 76319
IUPAC Name 2-methylbut-3-yn-2-amine
SMILES CC(C)(C#C)N

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.7900g/mL
Boiling Point 79.0°C to 80.0°C
Flash Point 2°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula HC≡CC(CH3)2NH2
Refractive Index 1.4220 to 1.4250
Quantity 25 g
Specific Gravity 0.79
Formula Weight 83.13
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 1, 1-Dimethylpropargylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,1-Dimethylpropargylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.