missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1-Dimethyloxirane, TRC
Click to view available options
Quantity:
50 g
Unit Size:
50g
Specifications
Specifications
| Chemical Name or Material | 1,1-Dimethyloxirane |
| CAS | 558-30-5 |
| Molecular Formula | C4H8O |
| Quantity | 50 g |
| Synonym | alpha,alpha-Dimethylethylene Oxide, 2-Epoxy-2-methyl-propane, 1,2-Epoxy-2-methylpropane, 1,2-Epoxyisobutane, 1,2-Isobutylene Oxide, 2,2-Dimethyloxirane, 2-Methyl-1,2-epoxypropane, 2-Methyl-1-propene oxide, 2-Methyl-1-propenoxide, 2-Methylpropylene oxide, Isobutene Oxide, Isobutylene Epoxide, Isobutylene Oxide, NSC 24249 |
| InChI Formula | InChI=1S/C4H8O/c1-4(2)3-5-4/h3H2,1-2H3 |
| SMILES | CC1(C)CO1 |
| IUPAC Name | 2,2-dimethyloxirane |
| Molecular Weight (g/mol) | 72.11 |
| Formula Weight | 72.0575 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction