missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1-Dimethylethyl Nitrite (~90%), TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | 1,1-Dimethylethyl Nitrite |
| CAS | 540-80-7 |
| Purity Grade Notes | GC |
| Molecular Formula | C4 H9 N O2 |
| Quantity | 10 g |
| Synonym | Nitrous acid, 1,1-dimethylethyl ester, Nitrous acid, tert-butyl ester (8CI), tert-Butyl nitrite (6CI,7CI), 1,1-Dimethylethyl nitrite, α,α-Dimethylethyl nitrite, Nitrous acid 1,1-dimethylethyl ester, Nitrous acid tert-butyl ester, tert-Butyl nitrite |
| InChI Formula | InChI=1S/C4H9NO2/c1-4(2,3)7-5-6/h1-3H3 |
| SMILES | CC(C)(C)ON=O |
| IUPAC Name | tert-butyl nitrite |
| Molecular Weight (g/mol) | 103.12 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction