missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dimethylethyl Nitrite (~90%), TRC

Product Code. 30384196
Click to view available options
Quantity:
25 g
Unit Size:
25g
This item is not returnable. View return policy

Product Code. 30384196

Brand: Toronto Research Chemicals TRCD47473025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,1-Dimethylethyl Nitrite
CAS 540-80-7
Purity Grade Notes GC
Molecular Formula C4 H9 N O2
Quantity 25 g
Synonym Nitrous acid, 1,1-dimethylethyl ester, Nitrous acid, tert-butyl ester (8CI), tert-Butyl nitrite (6CI,7CI), 1,1-Dimethylethyl nitrite, α,α-Dimethylethyl nitrite, Nitrous acid 1,1-dimethylethyl ester, Nitrous acid tert-butyl ester, tert-Butyl nitrite
InChI Formula InChI=1S/C4H9NO2/c1-4(2,3)7-5-6/h1-3H3
SMILES CC(C)(C)ON=O
IUPAC Name tert-butyl nitrite
Molecular Weight (g/mol) 103.12
Formula Weight 103.0633
Percent Purity >95
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.