missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dimethylethyl Ester N-(3-Iodopropyl)carbamic Acid, TRC

Product Code. 30357400
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357400 100 mg 100mg
30362566 250 mg -
30351877 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30357400 Supplier Toronto Research Chemicals Supplier No. TRCC183613100MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 167479-01-8
Molecular Formula C8H16NO2I
Molecular Weight (g/mol) 285.12
Synonym 1,1-Dimethylethyl N-(3-iodopropyl)carbamate, 1,1-Dimethylethyl (3-Iodopropyl)carbamate, tert-Butyl (3-Iodopropyl)carbamate, tert-Butyl N-(3-Iodopropyl)carbamate
IUPAC Name tert-butyl N-(3-iodopropyl)carbamate
SMILES CC(C)(C)OC(=O)NCCCI

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C8H16INO2/c1-8(2,3)12-7(11)10-6-4-5-9/h4-6H2,1-3H3,(H,10,11)
Formula Weight 285.0226
Chemical Name or Material 1,1-Dimethylethyl Ester N-(3-Iodopropyl)carbamic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.