missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dimethylethyl (4R-cis)-6-Aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate, Mikromol™

Product Code. 17900436
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity unitSize
17900436 100 mg 100mg
1 options
This item is not returnable. View return policy

Product Code. 17900436

Brand: Mikromol MM0400.05

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

1,1-Dimethylethyl (4R-cis)-6-Aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,1-Dimethylethyl (4R-cis)-6-Aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate
CAS 125995-13-3
Analyte or Component Names 1,1-Dimethylethyl (4R-cis)-6-Aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate
Shipping Condition Ice pack (-20°C)
Molecular Formula C14 H27 N O4
Certifications/Compliance ISO 9001
Synonym 1,1-Dimethylethyl (4R-cis)-6-Aminoethyl-2,2-dimethyl-1,3-dioxane-4-acetate
InChI Formula InChI=1S/C14H27NO4/c1-13(2,3)19-12(16)9-11-8-10(6-7-15)17-14(4,5)18-11/h10-11H,6-9,15H2,1-5H3/t10-,11-/m1/s1
SMILES CC(C)(C)OC(=O)C[C@H]1C[C@@H](CCN)OC(C)(C)O1
IUPAC Name tert-butyl 2-[(4R,6R)-6-(2-aminoethyl)-2,2-dimethyl-1,3-dioxan-4-yl]acetate
Molecular Weight (g/mol) 273.37
Quantity 100 mg
Formula Weight 273.194 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.