missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dimethylethyl 4'-(Bromomethyl)biphenyl-2-carboxylate, Mikromol™

Product Code. 30049648
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30049648 100 mg 100mg
30051456 25 mg 25mg
30461280 4 x 25 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30049648 Supplier Mikromol Supplier No. MM0939.08
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

1,1-Dimethylethyl 4'-(Bromomethyl)biphenyl-2-carboxylate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,1-Dimethylethyl 4'-(Bromomethyl)biphenyl-2-carboxylate
CAS 114772-40-6
Analyte or Component Names 1,1-Dimethylethyl 4'-(Bromomethyl)biphenyl-2-carboxylate
Shipping Condition Room Temperature
Molecular Formula C18 H19 Br O2
Certifications/Compliance ISO 9001
Synonym 1,1-Dimethylethyl 4'-(Bromomethyl)-biphenyl-2-carboxylate, 1,1-Dimethylethyl 4'-(bromomethyl)biphenyl-2-carboxylate, 2-(4-Bromomethylphenyl)benzoic acid tert-butyl ester, 2'-(tert-Butoxylcarbonyl)biphenyl-4-ylmethyl bromide, 4-(Bromomethyl)-2'-(tert-butoxycarbonyl)biphenyl, 4'-(Bromomethyl)-2-(tert-butoxycarbonyl)biphenyl, 4'-(Bromomethyl)[1,1'-biphenyl]-2-carboxylic acid 1,1-dimethylethyl ester, 4'-(Bromomethyl)biphenyl-2-carboxylic acid tert-butyl ester, 4'-Bromomethyl[1,1'-biphenyl]-2-carboxylic acid tert-butyl ester, 4'-Bromomethylbiphenyl-2-carboxylic Acid tert-Butyl Ester, tert-Butyl 2-(4-bromomethylphenyl)benzoate, tert-Butyl 4-(bromomethyl)biphenyl-2'-carboxylate, tert-Butyl 4-bromomethylbiphenyl-2-carboxylate, tert-Butyl 4'-(bromomethyl)biphenyl-2-carboxylate, Telmisartan Imp H (EP), Telmisartan Imp. H (EP), Telmisartan Impurity H
InChI Formula InChI=1S/C18H19BrO2/c1-18(2,3)21-17(20)16-7-5-4-6-15(16)14-10-8-13(12-19)9-11-14/h4-11H,12H2,1-3H3
SMILES CC(C)(C)OC(=O)c1ccccc1c2ccc(CBr)cc2
IUPAC Name tert-butyl 2-[4-(bromomethyl)phenyl]benzoate
Molecular Weight (g/mol) 347.25
Quantity 100 mg
Formula Weight 346.0568 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.