missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1-Dimethylethyl 4'-[[4-methyl-6-(1-methyl-1H-benzimidazol-2-yl)-2-propyl-1H-benzimidazol-1-yl]methyl]biphenyl-2-carboxylate (Telmisartan tert-Butyl Ester), Mikromol™
Description
1,1-Dimethylethyl 4'-[[4-methyl-6-(1-methyl-1H-benzimidazol-2-yl)-2-propyl-1H-benzimidazol-1-yl]methyl]biphenyl-2-carboxylate (Telmisartan tert-Butyl Ester)
Specifications
Specifications
| Chemical Name or Material | Telmisartan tert-Butyl Ester |
| CAS | 144702-26-1 |
| Analyte or Component Names | Telmisartan tert-Butyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C37 H38 N4 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1,1-Dimethylethyl 4'-[(1,4'-dimethyl-2'-propyl[2,6'-bi-1H-benzimidazol]-1'-yl)methyl][1,1'-biphenyl]-2-carboxylate, [1,1'-Biphenyl]-2-carboxylic acid, 4'-[(1,4'-dimethyl-2'-propyl[2,6'-bi-1H-benzimidazol]-1'-yl)methyl]-, 1,1-dimethylethyl ester, Telmisartan tert-Butyl Ester, Telmisartan Imp C (EP), Telmisartan Impurity C |
| InChI Formula | InChI=1S/C37H38N4O2/c1-7-12-33-39-34-24(2)21-27(35-38-30-15-10-11-16-31(30)40(35)6)22-32(34)41(33)23-25-17-19-26(20-18-25)28-13-8-9-14-29(28)36(42)43-37(3,4)5/h8-11,13-22H,7,12,23H2,1-6H3 |
| SMILES | CCCc1nc2c(C)cc(cc2n1Cc3ccc(cc3)c4ccccc4C(=O)OC(C)(C)C)c5nc6ccccc6n5C |
| IUPAC Name | tert-butyl 2-[4-[[4-methyl-6-(1-methylbenzimidazol-2-yl)-2-propylbenzimidazol-1-yl]methyl]phenyl]benzoate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?