missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dimethoxy-N,N-dimethylmethanamine, TRC

Product Code. 30381048
Change view
Click to view available options
Quantity:
5 g
Product Code. Quantity
30381048 5 g
1 options
This item is not returnable. View return policy

Product Code. 30381048

Brand: Toronto Research Chemicals TRCD4466255G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4637-24-5
Molecular Formula C5H13NO2
Quantity 5 g
Synonym 1,1-Dimethoxy-trimethylamine, (Dimethoxymethyl)dimethylamine, (Dimethylamino)dimethoxymethane, (Dimethylamino)formaldehyde Dimethyl Acetal, 1,1-Dimethoxy-N,N-dimethylmethanamine, 1,1-Dimethoxytrimethylamine, DMF Dimethyl Acetal, DMFDMA, Dimethoxy(dimethylamino)methane, Dimethoxy-N,N-dimethylmethanamine, Dimethoxy-N,N-dimethylmethylamine, Dimethyl Dimethylformamide Acetal, Dimethylformamide Dimethyl Acetal, Methyl-8, N,N-Dimethyl-1,1-bis(methyloxy)methanamine, N,N-Dimethylformamide Dimethyl Acetal, N-(Dimethoxymethyl)-N,N-dimethylamine, N-(Dimethoxymethyl)dimethylamine, α,α-Dimethoxytrimethylamine
InChI Formula InChI=1S/C5H13NO2/c1-6(2)5(7-3)8-4/h5H,1-4H3
SMILES COC(OC)N(C)C
Molecular Weight (g/mol) 119.16
Formula Weight 119.0946
Chemical Name or Material 1,1-Dimethoxy-N,N-dimethylmethanamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.