missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Diisooctyl Ester 2,2'-[(Dioctylstannylene)bis(thio)]bis-acetic Acid (Technical Grade), TRC

Product Code. 30384404
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30384404 1 g
30373778 500 mg
30377062 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30384404 Supplier Toronto Research Chemicals Supplier No. TRCD6388701G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26401-97-8
Molecular Formula C36H72O4S2Sn
Molecular Weight (g/mol) 751.79
Synonym Di-N-octyltin S,S-Bis(isooctyl thioglycolate), Dioctyltin S,S'-Bis(isooctyl mercaptoacetate)
IUPAC Name 6-methylheptyl 2-[[2-(6-methylheptoxy)-2-oxoethyl]sulfanyl-dioctylstannyl]sulfanylacetate
SMILES CCCCCCCC[Sn](CCCCCCCC)(SCC(=O)OCCCCCC(C)C)SCC(=O)OCCCCCC(C)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/2C10H20O2S.2C8H17.Sn/c2*1-9(2)6-4-3-5-7-12-10(11)8-13;2*1-3-5-7-8-6-4-2;/h2*9,13H,3-8H2,1-2H3;2*1,3-8H2,2H3;/q;;;;+2/p-2
Formula Weight 752.3894
Chemical Name or Material 1,1'-Diisooctyl Ester 2,2'-[(Dioctylstannylene)bis(thio)]bis-acetic Acid (Technical Grade)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.