missing translation for 'onlineSavingsMsg'
Learn More

1,1-Diethoxy-2-methyl-2-propanol, TRC

Product Code. 30379275
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30379275 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30379275 Supplier Toronto Research Chemicals Supplier No. TRCD442145250MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,1-Diethoxy-2-methyl-2-propanol
CAS 108994-55-4
Molecular Formula C25 H42 O4
Quantity 250 mg
Synonym 2-Methyl Lactaldehyde Diethyl Acetal
InChI Formula InChI=1S/C25H42O4/c1-15(5-10-23(28)29-4)19-8-9-20-18-7-6-16-13-17(26)11-12-24(16,2)21(18)14-22(27)25(19,20)3/h15-22,26-27H,5-14H2,1-4H3/t15-,16+,17-,18?,19+,20+,21?,22+,24-,25+/m0/s1
SMILES COC(=O)CC[C@H](C)[C@H]1CC[C@@H]2C3CC[C@@H]4C[C@@H](O)CC[C@]4(C)C3C[C@@H](O)[C@]12C
IUPAC Name methyl (4S)-4-[(3S,5R,10S,12R,13R,14R,17R)-3,12-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate
Molecular Weight (g/mol) 406.599
Formula Weight 406.308
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.