missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dichloroethene (stabilized with MEHQ), TRC

Product Code. 30387452
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387452 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30387452 Supplier Toronto Research Chemicals Supplier No. TRCD434360100ML

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 75-35-4
Molecular Formula C2 H2 Cl2
Quantity 100 mL
Synonym Ethylene, 1,1-dichloro- (8CI), 1,1-Dichloroethene, 1,1-Dichloroethylene, Diofan A 565S, F 1130a, HCC 1130a, Iso-dichloroethylene, R 1130a, Vinylidene chloride, XAN 10
InChI Formula InChI=1S/C2H2Cl2/c1-2(3)4/h1H2
SMILES ClC(=C)Cl
IUPAC Name 1,1-dichloroethene
Molecular Weight (g/mol) 96.94
Formula Weight 95.9534
Percent Purity >95%
Chemical Name or Material 1,1-Dichloroethene
Grade GC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.