missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dibromoethane, TRC

Product Code. 30381212
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Product Code. Quantity unitSize
30381212 500 mg 500mg
1 options
This item is not returnable. View return policy

Product Code. 30381212

Brand: Toronto Research Chemicals TRCD468205500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,1-Dibromoethane
CAS 557-91-5
Molecular Formula C2H4Br2
Quantity 500 mg
Synonym Ethylidene Dibromide
InChI Formula InChI=1S/C2H4Br2/c1-2(3)4/h2H,1H3
SMILES BrC(C)Br
Molecular Weight (g/mol) 187.86
Formula Weight 185.868
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.