missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dibromoethane, 99%, stabilized

Product Code. 10470641 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10470641 100 g Glass bottle 100g
10147510 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10470641 Supplier Thermo Scientific Acros Supplier No. 406481000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 557-91-5 | C2H4Br2 | 187.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 557-91-5
Molecular Formula C2H4Br2
Molecular Weight (g/mol) 187.86
MDL Number MFCD00041719
InChI Key APQIUTYORBAGEZ-UHFFFAOYSA-N
Synonym ethylidene bromide, ethane, 1,1-dibromo, ethylidene dibromide, unii-kj8zjy72qq, kj8zjy72qq, dibromethane, dibromoethan, dibromoetane, dibromo ethane, ccris 8552
PubChem CID 11201
IUPAC Name 1,1-dibromoethane
SMILES CC(Br)Br

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Brown
Density 2.0600g/mL
Boiling Point 107.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CHBr2
Refractive Index 1.5090 to 1.5120
Quantity 100 g
Specific Gravity 2.06
Stabilizer potassium carbonate
Formula Weight 187.86
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1,1-Dibromoethane, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,1-Dibromoethane, stabilized
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P313-Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.