missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dibromo-3,3,3-trifluoroacetone, 95%

Product Code. 11383006
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11383006 25 g 25g
11373006 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11383006 Supplier Thermo Scientific Alfa Aesar Supplier No. L10826.14
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 431-67-4 | C3HBr2F3O | 269.843 g/mol

1,1-Dibromo-3,3,3-trifluoroacetone readily forms trifluoropyruvaldehyde hydrate in the presence of aqueous NaOAc, which is a useful intermediate, e.g. in the synthesis of a fused pyrazine derivative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1-Dibromo-3,3,3-trifluoroacetone readily forms trifluoropyruvaldehyde hydrate in the presence of aqueous NaOAc, which is a useful intermediate, e.g. in the synthesis of a fused pyrazine derivative.

Solubility
Soluble in Chloroform. Not miscible or difficult to mix in water.

Notes
Keep container tightly closed. Incompatible with strong bases, active metals and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 431-67-4
Molecular Formula C3HBr2F3O
Molecular Weight (g/mol) 269.843
MDL Number MFCD00041362
InChI Key HEPPAPZASXFWTB-UHFFFAOYSA-N
Synonym 1,1-dibromo-3,3,3-trifluoroacetone, 3,3-dibromo-1,1,1-trifluoroacetone, 3,3-dibromo-1,1,1-trifluoro-propan-2-one, 2-propanone, 3,3-dibromo-1,1,1-trifluoro, dibromotrifluoroaceton, pubchem12627, 3,3-dibromotrifluoroacetone, ksc237m8t, l,l-dibromo-3,3,3-trifluoroacetone, 1,1-di-bromo-3,3,3-trifluoroacetone
PubChem CID 550523
IUPAC Name 3,3-dibromo-1,1,1-trifluoropropan-2-one
SMILES C(C(=O)C(F)(F)F)(Br)Br

Specifications

Boiling Point 111°C to 113°C
Odor Pungent
Refractive Index 1.4305
Quantity 25 g
UN Number UN3265
Beilstein 636645
Solubility Information Soluble in Chloroform. Not miscible or difficult to mix in water.
Formula Weight 269.85
Percent Purity 95%
Chemical Name or Material 1,1-Dibromo-3,3,3-trifluoroacetone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.