missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dibromo-3,3,3-trifluoroacetone, 95%

Product Code. 11373006
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11373006

Brand: Thermo Scientific Alfa Aesar L10826.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 431-67-4 | C3HBr2F3O | 269.843 g/mol

1,1-Dibromo-3,3,3-trifluoroacetone readily forms trifluoropyruvaldehyde hydrate in the presence of aqueous NaOAc, which is a useful intermediate, e.g. in the synthesis of a fused pyrazine derivative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1-Dibromo-3,3,3-trifluoroacetone readily forms trifluoropyruvaldehyde hydrate in the presence of aqueous NaOAc, which is a useful intermediate, e.g. in the synthesis of a fused pyrazine derivative.

Solubility
Soluble in Chloroform. Not miscible or difficult to mix in water.

Notes
Keep container tightly closed. Incompatible with strong bases, active metals and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 431-67-4
Molecular Formula C3HBr2F3O
Molecular Weight (g/mol) 269.843
MDL Number MFCD00041362
InChI Key HEPPAPZASXFWTB-UHFFFAOYSA-N
Synonym 1,1-dibromo-3,3,3-trifluoroacetone, 3,3-dibromo-1,1,1-trifluoroacetone, 3,3-dibromo-1,1,1-trifluoro-propan-2-one, 2-propanone, 3,3-dibromo-1,1,1-trifluoro, dibromotrifluoroaceton, pubchem12627, 3,3-dibromotrifluoroacetone, ksc237m8t, l,l-dibromo-3,3,3-trifluoroacetone, 1,1-di-bromo-3,3,3-trifluoroacetone
PubChem CID 550523
IUPAC Name 3,3-dibromo-1,1,1-trifluoropropan-2-one
SMILES C(C(=O)C(F)(F)F)(Br)Br

Specifications

Boiling Point 111°C to 113°C
Odor Pungent
Refractive Index 1.4305
Quantity 5 g
UN Number UN3265
Beilstein 636645
Solubility Information Soluble in Chloroform. Not miscible or difficult to mix in water.
Formula Weight 269.85
Percent Purity 95%
Chemical Name or Material 1,1-Dibromo-3,3,3-trifluoroacetone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.