missing translation for 'onlineSavingsMsg'
Learn More

1-(1-Cyclohexen-1-yl)piperidine, 97%

Product Code. 11360004
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11360004 5 g 5g
11370004 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11360004 Supplier Thermo Scientific Alfa Aesar Supplier No. L00335.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2981-10-4 | C8H22N2O2 | 178.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2981-10-4
MDL Number MFCD00014643

Specifications

Boiling Point 120°C to 122°C (16 mmHg)
Molecular Formula C8H22N2O2
Refractive Index 1.514
Quantity 5 g
Beilstein 113990
Synonym di 3-aminopropoxy ethane, 1-propanamine, 3,3'-1,2-ethanediylbis oxy bis, 1,2-bis 3-aminopropoxy ethane, 3,3'-ethane-1,2-diylbis oxy bis propan-1-amine, 3,3'-ethylenedioxy bispropylamine, 3,3'-ethylenedioxybis propylamine, propylamine, 3,3'-ethylenedioxy bis, ethylene glycol bis 3-aminopropyl ether, 1,10-diamino-4,7-dioxadecane, 3-2-3-aminopropoxy ethoxy propan-1-amine
InChI Key POTQBGGWSWSMCX-UHFFFAOYSA-P
SMILES C(CN)COCCOCCCN
IUPAC Name 3-[2-(3-aminopropoxy)ethoxy]propan-1-amine
Molecular Weight (g/mol) 178.28
PubChem CID 76342
Formula Weight 165.28
Percent Purity 97%
Chemical Name or Material 1-(1-Cyclohexen-1-yl)piperidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.